2-Methyl-5-(thien-2-yl)-3-furoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Jedi2, 2-Methyl-5-(thiophen-2-yl)furan-3-carboxylic acid

OR100020-1G OR100020-250MG
APOSOR100020-1GEA 272 EUR
APOSOR100020-1G APOSOR100020-250MG
2-Methyl-5-(thien-2-yl)-3-furoic acid
2-Methyl-5-(thien-2-yl)-3-furoic acid
Formel: C₁₀H₈O₃S
Molecular Weight: 208,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01569561
CAS-Nummer: 651005-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR