2-[(4-Chlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Carboxy-2-[(4-chlorophenyl)carbamoyl]cyclohexane, 2-Carboxy-N-(4-chlorophenyl)cyclohexane-1-carboxamide

OR10251-250MG OR10251-5G
APOSOR10251-250MGEA 74.1 EUR
APOSOR10251-250MG APOSOR10251-5G
2-[(4-Chlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
2-[(4-Chlorophenyl)carbamoyl]cyclohexane-1-carboxylic acid
Formel: C₁₄H₁₆ClNO₃
Molecular Weight: 281,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00590283
CAS-Nummer: 101937-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR