4-Aminotetrahydro-2H-pyran acetate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Tetrahydro-2H-pyran-4-amine acetate, 4-Aminooxane acetate

OR10342S-1G OR10342S-25G OR10342S-5G
APOSOR10342S-1GEA 21.9 EUR
APOSOR10342S-1G APOSOR10342S-25G APOSOR10342S-5G
4-Aminotetrahydro-2H-pyran acetate 98%
4-Aminotetrahydro-2H-pyran acetate
Formel: C₅H₁₁NO·CH₃CO₂H
Molecular Weight: 161.199 g/mol
Schmelzpunkt: 135…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445669
CAS-Nummer: 1005498-91-8
UN: 1325
ADR: 4.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR