3-Benzoyl-1-[(4-methylphenyl)sulphonyl]pyrrole 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Benzoyl-1-tosylpyrrole

OR10350-1G OR10350-5G OR10350-25G
APOSOR10350-1GEA 72.5 EUR
APOSOR10350-1G APOSOR10350-5G APOSOR10350-25G
3-Benzoyl-1-[(4-methylphenyl)sulphonyl]pyrrole 96%
3-Benzoyl-1-[(4-methylphenyl)sulphonyl]pyrrole
Formel: C₁₈H₁₅NO₃S
Molecular Weight: 325,382 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411960
CAS-Nummer: 139261-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR