Phenyl(1H-pyrrol-3-yl)methanone

Lieferant: Apollo Scientific
Achtung

Synonyme: [(1H-Pyrrol-3-yl)carbonyl]benzene, 3-Benzoyl-1H-pyrrole

OR10351-1G OR10351-5G
APOSOR10351-1GEA 183 EUR
APOSOR10351-1G APOSOR10351-5G
Phenyl(1H-pyrrol-3-yl)methanone
Phenyl(1H-pyrrol-3-yl)methanone
Formel: C₁₁H₉NO
Molecular Weight: 171,195 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179582
CAS-Nummer: 7126-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR