4-Cyanothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carboxy-4-cyanothiophene, 4-Cyano-2-thenoic acid, 5-Carboxythiophene-3-carbonitrile
OR103516-250MG
OR103516-1G
APOSOR103516-250MGEA
116
EUR
APOSOR103516-250MG
APOSOR103516-1G
4-Cyanothiophene-2-carboxylic acid
4-Cyanothiophene-2-carboxylic acid
Formel:
C₆H₃NO₂S Molecular Weight: 153,159 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664844 CAS-Nummer: 406719-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...