N,N-Dimethyl-4-iodo-1H-imidazole-1-sulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodo-N,N-dimethyl-1H-imidazole-1-sulphonamide

OR10456-1G OR10456-5G
APOSOR10456-1GEA 75.4 EUR
APOSOR10456-1G APOSOR10456-5G
N,N-Dimethyl-4-iodo-1H-imidazole-1-sulphonamide
N,N-Dimethyl-4-iodo-1H-imidazole-1-sulphonamide
Formel: C₅H₈IN₃O₂S
Molecular Weight: 301,105 g/mol
Schmelzpunkt: 130…133 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179543
CAS-Nummer: 135773-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR