3-[(E)-3-Methoxy-3-oxoprop-1-en-1-yl]benzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-boronocinnamate

OR10530-1G OR10530-5G
APOSOR10530-1GEA 183 EUR
APOSOR10530-1G APOSOR10530-5G
3-[(E)-3-Methoxy-3-oxoprop-1-en-1-yl]benzeneboronic acid 95%
3-[(E)-3-Methoxy-3-oxoprop-1-en-1-yl]benzeneboronic acid
Formel: C₁₀H₁₁BO₄
Molecular Weight: 206.003 g/mol
Schmelzpunkt: 126…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179477
CAS-Nummer: 380430-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR