(1E)-(Pent-1-en-1-yl)boronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(E)-Pent-1-en-1-ylboronic acid, trans-(Pent-1-en-1-yl)boronic acid
OR10576-1G
OR10576-25G
OR10576-5G
APOSOR10576-1GEA
105
EUR
APOSOR10576-1G
APOSOR10576-25G
APOSOR10576-5G
(1E)-(Pent-1-en-1-yl)boronic acid
(1E)-(Pent-1-en-1-yl)boronic acid
Formel:
C₅H₁₁BO₂ Molecular Weight: 113,951 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01074677 CAS-Nummer: 59239-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...