[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide

Lieferant: Apollo Scientific
Achtung

Synonyme: [4-Ethoxy-4-oxo(but-1-yl)]tris(phenyl)phosphonium bromide, (4-Ethoxy-4-oxobutyl)triphenylphosphonium bromide

OR10652-25G
APOSOR10652-25GEA 72.5 EUR
APOSOR10652-25G
[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide
[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide
Formel: C₂₄H₂₆O₂P·Br
Molecular Weight: 457,34 g/mol
Schmelzpunkt: 165…167 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00274610
CAS-Nummer: 50479-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR