2-(4-Hydroxyphenyl)-N,N-dimethylacetamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[2-(Dimethylamino)-2-oxoethyl]phenol, 4-[(Dimethylcarbamoyl)methyl]phenol

OR10858-1G OR10858-5G
APOSOR10858-1GEA 319 EUR
APOSOR10858-1G APOSOR10858-5G
2-(4-Hydroxyphenyl)-N,N-dimethylacetamide
2-(4-Hydroxyphenyl)-N,N-dimethylacetamide
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03453072
CAS-Nummer: 76472-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR