4-[(Methylsulphonyl)amino]benzoic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-(4-Carboxyphenyl)methanesulphonamide, 4-(Methanesulphonamido)benzoic acid, 4-(Methylsulphonamido)benzoic acid

OR10892-1G OR10892-250MG OR10892-5G
APOSOR10892-1GEA 49.3 EUR
APOSOR10892-1G APOSOR10892-250MG APOSOR10892-5G
4-[(Methylsulphonyl)amino]benzoic acid
4-[(Methylsulphonyl)amino]benzoic acid
Formel: C₈H₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00025052
CAS-Nummer: 7151-76-0
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR