Ethyl 2-ethoxynicotinate

Lieferant: Apollo Scientific

OR110041-1G OR110041-5G OR110041-10G
APOSOR110041-1GEA 116 EUR
APOSOR110041-1G APOSOR110041-5G APOSOR110041-10G
Ethyl 2-ethoxynicotinate
Ethyl 2-ethoxynicotinate
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18207652
CAS-Nummer: 15441-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR