5-Nitro-2,3-dihydro-1-benzofuran

Lieferant: Apollo Scientific

Synonyme: 5-Nitro-2,3-dihydrobenzofuran

OR110049-5G OR110049-10G
APOSOR110049-5GEA 31.9 EUR
APOSOR110049-5G APOSOR110049-10G
5-Nitro-2,3-dihydro-1-benzofuran
5-Nitro-2,3-dihydro-1-benzofuran
Formel: C₈H₇NO₃
Molecular Weight: 165.146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22376562
CAS-Nummer: 17403-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR