5,7-Dibromo-4-nitro-1H-indazole, tech
Lieferant: Apollo Scientific
|
Achtung
|
OR110061-1G
OR110061-10G
OR110061-5G
APOSOR110061-1GEA
0
EUR
APOSOR110061-1G
APOSOR110061-10G
APOSOR110061-5G
5,7-Dibromo-4-nitro-1H-indazole, tech
5,7-Dibromo-4-nitro-1H-indazole
Formel:
C₇H₃Br₂N₃O₂ Molecular Weight: 320,926 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23159157 CAS-Nummer: 1427460-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...