4,5-Dichloro-6-methyl-2-(pyridin-4-yl)pyrimidine

Lieferant: Apollo Scientific

OR110075-1G OR110075-5G
APOSOR110075-1GEA 296 EUR
APOSOR110075-1G APOSOR110075-5G
4,5-Dichloro-6-methyl-2-(pyridin-4-yl)pyrimidine
4,5-Dichloro-6-methyl-2-(pyridin-4-yl)pyrimidine
Formel: C₁₀H₇Cl₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00126821
CAS-Nummer: 1239850-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR