6-Amino-1-heptyl-1H-indazole

Lieferant: Apollo Scientific

Synonyme: 1-Heptyl-1H-indazol-6-amine

OR110115-1G OR110115-250MG
APOSOR110115-1GEA 372 EUR
APOSOR110115-1G APOSOR110115-250MG
6-Amino-1-heptyl-1H-indazole
6-Amino-1-heptyl-1H-indazole
Formel: C₁₄H₂₁N₃
Molecular Weight: 231,337 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD12103247
CAS-Nummer: 1155102-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR