1,3-Benzoxazole-6-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[d]oxazole-6-carboxylic acid

OR110125-5G
APOSOR110125-5GEA 82.8 EUR
APOSOR110125-5G
1,3-Benzoxazole-6-carboxylic acid 97%
1,3-Benzoxazole-6-carboxylic acid
Formel: C₈H₅NO₃
Molecular Weight: 163,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954949
CAS-Nummer: 154235-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR