Methyl 7-bromo-1H-indazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110164-1G OR110164-250MG
APOSOR110164-1GEA 277 EUR
APOSOR110164-1G APOSOR110164-250MG
Methyl 7-bromo-1H-indazole-5-carboxylate
Methyl 7-bromo-1H-indazole-5-carboxylate
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,068 g/mol
Schmelzpunkt: 205…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22421936
CAS-Nummer: 1427460-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR