4-Chloro-N-(1-methyl-1H-indazol-3-yl)butanamide

Lieferant: Apollo Scientific

OR110171-1G OR110171-250MG
APOSOR110171-1GEA 232 EUR
APOSOR110171-1G APOSOR110171-250MG
4-Chloro-N-(1-methyl-1H-indazol-3-yl)butanamide
4-Chloro-N-(1-methyl-1H-indazol-3-yl)butanamide
Formel: C₁₂H₁₄ClN₃O
Molecular Weight: 251,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22370026
CAS-Nummer: 1427460-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR