Ethyl (2-methyl-1,3-thiazol-4-yl)acetate
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(2-methyl-1,-3-thiazol-4-yl)-acetate
OR110217-5G
OR110217-25G
APOSOR110217-5GEA
125
EUR
APOSOR110217-5G
APOSOR110217-25G
Ethyl (2-methyl-1,3-thiazol-4-yl)acetate
Ethyl (2-methyl-1,3-thiazol-4-yl)acetate
Formel:
C₈H₁₁NO₂S Molecular Weight: 185,243 g/mol Siedepunkt: 114 °C Dichte: 1,174 g/cm³ (25 °C) Flammpunkt: 110,4 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052519 CAS-Nummer: 37128-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...