5,6-Dichloro-2-ethyl-1H-benzimidazole

Lieferant: Apollo Scientific

OR110225-1G OR110225-5G
APOSOR110225-1GEA 0 EUR
APOSOR110225-1G APOSOR110225-5G
5,6-Dichloro-2-ethyl-1H-benzimidazole
5,6-Dichloro-2-ethyl-1H-benzimidazole
Formel: C₉H₈Cl₂N₂
Molecular Weight: 215,079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05992934
CAS-Nummer: 90348-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR