Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-8-carboxylate

OR110233-1G OR110233-5G
APOSOR110233-1GEA 177 EUR
APOSOR110233-1G APOSOR110233-5G
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-8-carboxylate
Formel: C₁₀H₉NO₄
Schmelzpunkt: 203…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02930634
CAS-Nummer: 149396-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR