2-(4-Nitrophenyl)-5-propyl-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR110237-1G OR110237-250MG
APOSOR110237-1GEA 206 EUR
APOSOR110237-1G APOSOR110237-250MG
2-(4-Nitrophenyl)-5-propyl-1,3,4-oxadiazole
2-(4-Nitrophenyl)-5-propyl-1,3,4-oxadiazole
Formel: C₁₁H₁₁N₃O₃
Molecular Weight: 233.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11094979
CAS-Nummer: 944661-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR