8-Bromo-5,5,7,7-tetramethyl-4-nitro-1,5,6,7-tetrahydrocyclopenta[f]indazole
Lieferant: Apollo Scientific
OR110241-1G
OR110241-250MG
APOSOR110241-1GEA
348
EUR
APOSOR110241-1G
APOSOR110241-250MG
8-Bromo-5,5,7,7-tetramethyl-4-nitro-1,5,6,7-tetrahydrocyclopenta[f]indazole
8-Bromo-5,5,7,7-tetramethyl-4-nitro-1,5,6,7-tetrahydrocyclopenta[f]indazole
Formel:
C₁₄H₁₆BrN₃O₂ Molecular Weight: 338,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846257 CAS-Nummer: 1227955-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...