4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific

OR110244-1G OR110244-250MG
APOSOR110244-1GEA 221 EUR
APOSOR110244-1G APOSOR110244-250MG
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₈N₂O₄
Molecular Weight: 208.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406815
CAS-Nummer: 132522-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR