6-Methoxy-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific

OR110245-1G OR110245-250MG
APOSOR110245-1GEA 206 EUR
APOSOR110245-1G APOSOR110245-250MG
6-Methoxy-2H-1,4-benzoxazin-3(4H)-one
6-Methoxy-2H-1,4-benzoxazin-3(4H)-one
Formel: C₉H₉NO₃
Molecular Weight: 179,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272060
CAS-Nummer: 5023-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR