Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-bromobenzo[b]thiophene-2-carboxylate, Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate

OR110258-10G OR110258-1G OR110258-5G
APOSOR110258-10GEA 400 EUR
APOSOR110258-10G APOSOR110258-1G APOSOR110258-5G
Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate
Ethyl 5-bromo-1-benzo[b]thiophene-2-carboxylate
Formel: C₁₁H₉BrO₂S
Molecular Weight: 285.157 g/mol
Schmelzpunkt: 86…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12756246
CAS-Nummer: 13771-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR