1-Acetyl-4-(4-nitrophenyl)homopiperazine

Lieferant: Apollo Scientific

Synonyme: 1-Acetyl-4-(4-nitrophenyl)-1,4-diazepane

OR110262-1G OR110262-250MG
APOSOR110262-1GEA 232 EUR
APOSOR110262-1G APOSOR110262-250MG
1-Acetyl-4-(4-nitrophenyl)homopiperazine
1-Acetyl-4-(4-nitrophenyl)homopiperazine
Formel: C₁₃H₁₇N₃O₃
Molecular Weight: 263.292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12964044
CAS-Nummer: 892491-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR