Methyl 2-butyl-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110271-1G OR110271-5G
APOSOR110271-1GEA 128 EUR
APOSOR110271-1G APOSOR110271-5G
Methyl 2-butyl-1,3-benzoxazole-5-carboxylate
Methyl 2-butyl-1,3-benzoxazole-5-carboxylate
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13182317
CAS-Nummer: 1305712-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR