N-Methylthiophene-2-sulphonamide

Lieferant: Apollo Scientific

OR110294-1G OR110294-250MG OR110294-5G
APOSOR110294-1GEA 203 EUR
APOSOR110294-1G APOSOR110294-250MG APOSOR110294-5G
N-Methylthiophene-2-sulphonamide
N-Methylthiophene-2-sulphonamide
Formel: C₅H₇NO₂S₂
Molecular Weight: 177.245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975951
CAS-Nummer: 53442-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR