4-(1,3-Thiazol-2-yl)pyrimidin-2-amine

Lieferant: Apollo Scientific
Achtung

OR110323-250MG
APOSOR110323-250MGEA 60.9 EUR
APOSOR110323-250MG
4-(1,3-Thiazol-2-yl)pyrimidin-2-amine
4-(1,3-Thiazol-2-yl)pyrimidin-2-amine
Formel: C₇H₆N₄S
Molecular Weight: 178,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00105074
CAS-Nummer: 874830-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR