2-Methyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

OR110334-1G OR110334-5G
APOSOR110334-1GEA 69.6 EUR
APOSOR110334-1G APOSOR110334-5G
2-Methyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
2-Methyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12755875
CAS-Nummer: 20348-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR