Imidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110423-1G OR110423-250MG OR110423-5G
APOSOR110423-1GEA 112 EUR
APOSOR110423-1G APOSOR110423-250MG APOSOR110423-5G
Imidazo[1,2-a]pyridine-2-carbohydrazide
Imidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₈N₄O
Molecular Weight: 176.175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012810
CAS-Nummer: 119448-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR