Ethyl 4-(cyanomethoxy)benzoate

Lieferant: Apollo Scientific

OR110464-1G OR110464-5G
APOSOR110464-1GEA 173 EUR
APOSOR110464-1G APOSOR110464-5G
Ethyl 4-(cyanomethoxy)benzoate
Ethyl 4-(cyanomethoxy)benzoate
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109536
CAS-Nummer: 501662-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR