[4-(Benzyloxy)phenoxy]acetonitrile

Lieferant: Apollo Scientific

OR110466-1G OR110466-5G
APOSOR110466-1GEA 223 EUR
APOSOR110466-1G APOSOR110466-5G
[4-(Benzyloxy)phenoxy]acetonitrile
[4-(Benzyloxy)phenoxy]acetonitrile
Formel: C₁₅H₁₃NO₂
Molecular Weight: 239,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09944824
CAS-Nummer: 50635-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR