4-(Oct-1-yl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: [4-(Oct-1-yl)phenyl]methanol

OR110470-1G OR110470-5G
APOSOR110470-1GEA 212 EUR
APOSOR110470-1G APOSOR110470-5G
4-(Oct-1-yl)benzyl alcohol
4-(Oct-1-yl)benzyl alcohol
Formel: C₁₅H₂₄O
Molecular Weight: 220.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202723
CAS-Nummer: 40016-25-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR