2-(3-tert-Butylphenoxy)benzaldehyde

Lieferant: Apollo Scientific

OR110501-1G OR110501-5G
APOSOR110501-1GEA 186 EUR
APOSOR110501-1G APOSOR110501-5G
2-(3-tert-Butylphenoxy)benzaldehyde
2-(3-tert-Butylphenoxy)benzaldehyde
Formel: C₁₇H₁₈O₂
Molecular Weight: 254,324 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12570767
CAS-Nummer: 1099155-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR