[2-(3-tert-Butylphenoxy)phenyl]methanol

Lieferant: Apollo Scientific

OR110508-1G OR110508-5G
APOSOR110508-1GEA 248 EUR
APOSOR110508-1G APOSOR110508-5G
[2-(3-tert-Butylphenoxy)phenyl]methanol
[2-(3-tert-Butylphenoxy)phenyl]methanol
Formel: C₁₇H₂₀O₂
Molecular Weight: 256.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12656401
CAS-Nummer: 1098366-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR