[4-(4-Methylphenoxy)phenyl]methanol

Lieferant: Apollo Scientific

OR110521-1G OR110521-5G
APOSOR110521-1GEA 248 EUR
APOSOR110521-1G APOSOR110521-5G
[4-(4-Methylphenoxy)phenyl]methanol
[4-(4-Methylphenoxy)phenyl]methanol
Formel: C₁₄H₁₄O₂
Molecular Weight: 214,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11190353
CAS-Nummer: 181231-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR