[4-(3-Methylphenoxy)phenyl]methanol
Lieferant: Apollo Scientific
OR110522-1G
OR110522-5G
APOSOR110522-1GEA
248
EUR
APOSOR110522-1G
APOSOR110522-5G
[4-(3-Methylphenoxy)phenyl]methanol
[4-(3-Methylphenoxy)phenyl]methanol
Formel:
C₁₄H₁₄O₂ Molecular Weight: 214,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11190414 CAS-Nummer: 53818-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...