Methyl 2'-chloro-1,1'-biphenyl-4-carboxylate

Lieferant: Apollo Scientific

OR110532-1G OR110532-5G
APOSOR110532-1GEA 296 EUR
APOSOR110532-1G APOSOR110532-5G
Methyl 2'-chloro-1,1'-biphenyl-4-carboxylate
Methyl 2'-chloro-1,1'-biphenyl-4-carboxylate
Formel: C₁₄H₁₁ClO₂
Molecular Weight: 246,689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05981681
CAS-Nummer: 89900-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR