4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene
Lieferant: Apollo Scientific
OR110535-1G
OR110535-5G
OR110535-10G
APOSOR110535-1GEA
186
EUR
APOSOR110535-1G
APOSOR110535-5G
APOSOR110535-10G
4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene
4-(4-Bromophenoxy)-2-methyl-1-nitrobenzene
Formel:
C₁₃H₁₀BrNO₃ Molecular Weight: 308,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12069426 CAS-Nummer: 163258-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...