2-(1H-Pyrazol-1-yl)isonicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(1H-Pyrazol-1-yl)pyridine-4-carboxylic acid

OR110593-500MG
APOSOR110593-500MGEA 183 EUR
APOSOR110593-500MG
2-(1H-Pyrazol-1-yl)isonicotinic acid
2-(1H-Pyrazol-1-yl)isonicotinic acid
Formel: C₉H₇N₃O₂
Molecular Weight: 189,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11128544
CAS-Nummer: 1152940-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR