3-(Dimethylamino)-1-pyridin-4-ylprop-2-en-1-one

Lieferant: Apollo Scientific

Synonyme: (E)-3-(Dimethylamino)-1-(pyridin-4-yl)prop-2-en-1-one

OR110599-10G OR110599-1G OR110599-5G
APOSOR110599-10GEA 741 EUR
APOSOR110599-10G APOSOR110599-1G APOSOR110599-5G
3-(Dimethylamino)-1-pyridin-4-ylprop-2-en-1-one
3-(Dimethylamino)-1-pyridin-4-ylprop-2-en-1-one
Formel: C₁₀H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00115179
CAS-Nummer: 123367-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR