1-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]ethanone

Lieferant: Apollo Scientific

OR110604-1G OR110604-10G OR110604-5G
APOSOR110604-1GEA 248 EUR
APOSOR110604-1G APOSOR110604-10G APOSOR110604-5G
1-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]ethanone
1-[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]ethanone
Formel: C₁₁H₉BrN₂O
Molecular Weight: 265,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756230
CAS-Nummer: 1182992-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR