2-Bromo-1-[4-(4-bromo-1H-pyrazol-1-yl)phenyl]ethanone

Lieferant: Apollo Scientific

OR110605-1G OR110605-5G
APOSOR110605-1GEA 296 EUR
APOSOR110605-1G APOSOR110605-5G
2-Bromo-1-[4-(4-bromo-1H-pyrazol-1-yl)phenyl]ethanone
2-Bromo-1-[4-(4-bromo-1H-pyrazol-1-yl)phenyl]ethanone
Formel: C₁₁H₈Br₂N₂O
Molecular Weight: 344,002 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756234
CAS-Nummer: 1227955-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR