tert-Butyl 4-oxo-3,5,6,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine-7(4H)-carboxylate

Lieferant: Apollo Scientific

OR110627-1G OR110627-5G
APOSOR110627-1GEA 261 EUR
APOSOR110627-1G APOSOR110627-5G
tert-Butyl 4-oxo-3,5,6,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine-7(4H)-carboxylate
tert-Butyl 4-oxo-3,5,6,8-tetrahydropyrido[4',3':4,5]thieno[2,3-d]pyrimidine-7(4H)-carboxylate
Formel: C₁₄H₁₇N₃O₃S
Molecular Weight: 307.368 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664943
CAS-Nummer: 874801-60-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR