5-Methylthiophene-2-sulphonamide
Lieferant: Apollo Scientific
|
Achtung
|
OR110640-10G
OR110640-1G
OR110640-5G
APOSOR110640-10GEA
445
EUR
APOSOR110640-10G
APOSOR110640-1G
APOSOR110640-5G
5-Methylthiophene-2-sulphonamide
5-Methylthiophene-2-sulphonamide
Formel:
C₅H₇NO₂S₂ Molecular Weight: 177,245 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08444612 CAS-Nummer: 53595-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...