2,4-Dibromo-6-tert-butylphenol
Lieferant: Apollo Scientific
Synonyme:
2,4-Dibromo-6-(tert-butyl)phenol
OR110661-1G
OR110661-5G
OR110661-10G
APOSOR110661-1GEA
198
EUR
APOSOR110661-1G
APOSOR110661-5G
APOSOR110661-10G
2,4-Dibromo-6-tert-butylphenol
2,4-Dibromo-6-tert-butylphenol
Formel:
C₁₀H₁₂Br₂O Molecular Weight: 308.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463268 CAS-Nummer: 15460-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...